siRNA Details
| Gene Symbol: LOC284661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027088.1 | 4 | 1067 | |||
| GenBank mRNA | AK093498.1 | - | 4 | 1067 | ||
| BC128213.1 | - | 3 | 479 | |||
| BC128214.1 | - | 3 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284661| Gene Symbol: LOC284661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027088.1 | 4 | 1067 | |||
| GenBank mRNA | AK093498.1 | - | 4 | 1067 | ||
| BC128213.1 | - | 3 | 479 | |||
| BC128214.1 | - | 3 | 480 | |||