siRNA Details
| Gene Symbol: LINC01142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026957.1 | 3 | 796 | |||
| GenBank mRNA | BC033381.2 | - | 2 | 726 | ||
| BC125144.1 | - | 1 | 193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01142| Gene Symbol: LINC01142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026957.1 | 3 | 796 | |||
| GenBank mRNA | BC033381.2 | - | 2 | 726 | ||
| BC125144.1 | - | 1 | 193 | |||