siRNA Details
| Gene Symbol: DPPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018189.3 | NP_060659.3 | 7 | 948 | ||
| XM_011512954.2 | XP_011511256.1 | 7 | 1227 | |||
| GenBank mRNA | AK001575.1 | - | 7 | 890 | ||
| AK022698.1 | - | 7 | 904 | |||
| AK022821.1 | - | 2 | 1867 | |||
| AK290992.1 | - | 7 | 941 | |||
| AK298623.1 | - | 6 | 602 | |||
| BC032846.2 | - | 7 | 922 | 5267235 | ||

DPPA4
Genomic Map