siRNA Details
Gene Symbol: LOC284798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027091.1 | 4 | 3102 | |||
NR_027092.1 | 4 | 2395 | ||||
NR_027093.1 | 4 | 2502 | ||||
GenBank mRNA | AK095800.1 | - | 4 | 3102 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027091.1 | 4 | 3102 | |||
NR_027092.1 | 4 | 2395 | ||||
NR_027093.1 | 4 | 2502 | ||||
GenBank mRNA | AK095800.1 | - | 4 | 3102 |