siRNA Details
Gene Symbol: LINC00692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034055.1 | 8 | 699 | |||
GenBank mRNA | BC033370.1 | - | 8 | 702 | ||
BC126324.1 | - | 7 | 536 | |||
BC143987.1 | - | 7 | 613 | |||
BC143998.1 | - | 7 | 610 | |||
BC143999.1 | - | 7 | 640 |
