siRNA Details
Gene Symbol: ZNF852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287349.1 | NP_001274278.1 | 3 | 206 | ||
GenBank mRNA | AK131507.1 | - | 1 | 468 | ||
AK296954.1 | - | 3 | 202 | |||
BC014381.1 | - | 2 | 92 | |||
DC398999.1 | - | 3 | 207 |
