siRNA Details
| Gene Symbol: LINC00698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027104.1 | 7 | 1690 | |||
| GenBank mRNA | BC039502.1 | - | 6 | 1282 | ||
| BC043407.1 | - | 9 | 1811 | |||
| BC104474.1 | - | 1 | 592 | |||
| BC104475.1 | - | 1 | 592 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00698| Gene Symbol: LINC00698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027104.1 | 7 | 1690 | |||
| GenBank mRNA | BC039502.1 | - | 6 | 1282 | ||
| BC043407.1 | - | 9 | 1811 | |||
| BC104474.1 | - | 1 | 592 | |||
| BC104475.1 | - | 1 | 592 | |||