siRNA Details
| Gene Symbol: F11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033901.2 | 4 | 831 | |||
| GenBank mRNA | BC038717.1 | - | 4 | 770 | ||
| BC117432.1 | - | 3 | 557 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11-AS1| Gene Symbol: F11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033901.2 | 4 | 831 | |||
| GenBank mRNA | BC038717.1 | - | 4 | 770 | ||
| BC117432.1 | - | 3 | 557 | |||