siRNA Details
| Gene Symbol: LINC01019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033898.1 | 4 | 930 | |||
| GenBank mRNA | BC035019.1 | - | 4 | 882 | ||
| BC110532.1 | - | 2 | 385 | |||
| BC122538.1 | - | 2 | 386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01019| Gene Symbol: LINC01019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033898.1 | 4 | 930 | |||
| GenBank mRNA | BC035019.1 | - | 4 | 882 | ||
| BC110532.1 | - | 2 | 385 | |||
| BC122538.1 | - | 2 | 386 | |||