siRNA Details
| Gene Symbol: NBPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011540763.1 | XP_011539065.1 | 16 | 2777 | ||
| Gene Symbol: NBPF22P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003719.2 | 8 | 2009 | |||
| GenBank mRNA | BC050328.1 | - | 8 | 1965 | ||

NBPF4
Genomic Map