siRNA Details
Gene Symbol: NBPF22P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003719.2 | 8 | 1498 | |||
GenBank mRNA | AK308274.1 | - | 8 | 1482 | ||
BC050328.1 | - | 8 | 1454 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NBPF22P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003719.2 | 8 | 1498 | |||
GenBank mRNA | AK308274.1 | - | 8 | 1482 | ||
BC050328.1 | - | 8 | 1454 |