siRNA Details
| Gene Symbol: LOC285626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037889.1 | 5 | 1148 | |||
| GenBank mRNA | AK097548.1 | - | 5 | 1147 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285626| Gene Symbol: LOC285626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037889.1 | 5 | 1148 | |||
| GenBank mRNA | AK097548.1 | - | 5 | 1147 | ||