siRNA Details
| Gene Symbol: LOC285638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833206.1 | - | 4 | 5433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285638| Gene Symbol: LOC285638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL833206.1 | - | 4 | 5433 | ||