siRNA Details
| Gene Symbol: CNPY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103176.1 | NP_001096646.1 | 4 | 607 | ||
| GenBank mRNA | AK094445.1 | - | 4 | 625 | ||
| BC105732.1 | - | 4 | 607 | |||
| BC105733.1 | - | 4 | 607 | |||
| BC105734.1 | - | 4 | 607 | |||
| BC107598.1 | - | 4 | 607 | |||

CNPY1
Genomic Map