siRNA Details
Gene Symbol: FAM183CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024473.1 | 3 | 2591 | |||
GenBank mRNA | AL833525.1 | - | 3 | 2285 | ||
BC028745.1 | - | 4 | 2122 | |||
BC045833.1 | - | 4 | 2117 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM183CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024473.1 | 3 | 2591 | |||
GenBank mRNA | AL833525.1 | - | 3 | 2285 | ||
BC028745.1 | - | 4 | 2122 | |||
BC045833.1 | - | 4 | 2117 |