siRNA Details
| Gene Symbol: SLC25A25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033374.1 | 4 | 1384 | |||
| GenBank mRNA | AK124723.1 | - | 4 | 1382 | ||
| AL833509.1 | - | 4 | 1343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A25-AS1| Gene Symbol: SLC25A25-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033374.1 | 4 | 1384 | |||
| GenBank mRNA | AK124723.1 | - | 4 | 1382 | ||
| AL833509.1 | - | 4 | 1343 | |||