siRNA Details
| Gene Symbol: FAM225A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024366.1 | 3 | 5309 | |||
| GenBank mRNA | AK127516.1 | - | 1 | 4295 | ||
| AL832752.1 | - | 3 | 5132 | |||
| BX647840.1 | - | 2 | 5536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM225A| Gene Symbol: FAM225A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024366.1 | 3 | 5309 | |||
| GenBank mRNA | AK127516.1 | - | 1 | 4295 | ||
| AL832752.1 | - | 3 | 5132 | |||
| BX647840.1 | - | 2 | 5536 | |||