siRNA Details
| Gene Symbol: FIRRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026975.1 | 8, 9 | 1637 | |||
| GenBank mRNA | BC131554.1 | - | 8, 9 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FIRRE| Gene Symbol: FIRRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026975.1 | 8, 9 | 1637 | |||
| GenBank mRNA | BC131554.1 | - | 8, 9 | 1093 | ||