siRNA Details
| Gene Symbol: ANKRD30BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043208.2 | - | 9, 10 | 2180 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD30BP3| Gene Symbol: ANKRD30BP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC043208.2 | - | 9, 10 | 2180 | ||