siRNA Details
| Gene Symbol: C17orf112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243552.1 | NP_001230481.1 | 4 | 652 | ||
| XM_017024004.1 | XP_016879493.1 | 4 | 589 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17orf112| Gene Symbol: C17orf112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243552.1 | NP_001230481.1 | 4 | 652 | ||
| XM_017024004.1 | XP_016879493.1 | 4 | 589 | |||