siRNA Details
| Gene Symbol: LOC105372444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136526.1 | 3 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372444| Gene Symbol: LOC105372444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136526.1 | 3 | 578 | |||