siRNA Details
| Gene Symbol: MROH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287395.1 | NP_001274324.1 | 22, 23 | 2558 | ||
| XM_011511079.2 | XP_011509381.1 | 22, 23 | 2735 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MROH2A| Gene Symbol: MROH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287395.1 | NP_001274324.1 | 22, 23 | 2558 | ||
| XM_011511079.2 | XP_011509381.1 | 22, 23 | 2735 | |||