siRNA Details
| Gene Symbol: KIAA2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277372.2 | NP_001264301.2 | 13 | 2249 | ||
| XM_006712172.2 | XP_006712235.1 | 13 | 2254 | |||
| XM_011510467.2 | XP_011508769.1 | 13 | 2362 | |||
| XM_011510468.2 | XP_011508770.1 | 13 | 2257 | |||
| XM_017003111.1 | XP_016858600.1 | 13 | 2362 | |||
| GenBank mRNA | AB095932.1 | - | 1 | 2757 | ||
| AK295603.1 | - | 15 | 2287 | |||
| AK297413.1 | - | 14 | 2439 | |||
| BC118982.1 | - | 1 | 570 | 40026312 | ||

KIAA2012
Genomic Map