siRNA Details
Gene Symbol: LOC101060498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121640.1 | 3 | 835 | |||
NR_121641.1 | 4 | 691 | ||||
GenBank mRNA | JN120857.1 | - | 3 | 583 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101060498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121640.1 | 3 | 835 | |||
NR_121641.1 | 4 | 691 | ||||
GenBank mRNA | JN120857.1 | - | 3 | 583 |