siRNA Details
| Gene Symbol: ESRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027122.1 | 3 | 2443 | |||
| GenBank mRNA | DQ445779.2 | - | 3 | 2442 | ||
| KM363598.1 | - | 3 | 2443 | |||
| Gene Symbol: LOC340107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037896.1 | 3 | 1807 | |||
| GenBank mRNA | BC040315.1 | - | 3 | 1807 | ||
| BC122542.1 | - | 1 | 271 | |||

ESRG
Genomic Map