siRNA Details
Gene Symbol: FAM183BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028347.1 | 2 | 1127 | |||
GenBank mRNA | AK124132.1 | - | 1 | 1224 | ||
BC045803.1 | - | 2 | 1075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM183BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028347.1 | 2 | 1127 | |||
GenBank mRNA | AK124132.1 | - | 1 | 1224 | ||
BC045803.1 | - | 2 | 1075 |