siRNA Details
| Gene Symbol: FAM183BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028347.1 | 2 | 1061 | |||
| GenBank mRNA | AK124132.1 | - | 1 | 1158 | ||
| BC045803.1 | - | 2 | 1009 | |||
| FJ211598.1 | - | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM183BP| Gene Symbol: FAM183BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028347.1 | 2 | 1061 | |||
| GenBank mRNA | AK124132.1 | - | 1 | 1158 | ||
| BC045803.1 | - | 2 | 1009 | |||
| FJ211598.1 | - | 1 | 74 | |||