siRNA Details
| Gene Symbol: SLFN12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017024005.1 | XP_016879494.1 | 2, 3 | 976 | ||
| GenBank mRNA | AK172761.1 | - | 2, 3 | 811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLFN12L| Gene Symbol: SLFN12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017024005.1 | XP_016879494.1 | 2, 3 | 976 | ||
| GenBank mRNA | AK172761.1 | - | 2, 3 | 811 | ||