siRNA Details
Gene Symbol: NOTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134462.1 | NP_001127934.1 | 3 | 1035 | ||
GenBank mRNA | AK295936.1 | - | 3 | 1035 | ||
AK316410.1 | - | 3 | 991 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NOTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134462.1 | NP_001127934.1 | 3 | 1035 | ||
GenBank mRNA | AK295936.1 | - | 3 | 1035 | ||
AK316410.1 | - | 3 | 991 |