siRNA Details
| Gene Symbol: GBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136680.1 | - | 5, 6 | 833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GBP3| Gene Symbol: GBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136680.1 | - | 5, 6 | 833 | ||