siRNA Details
| Gene Symbol: LRRIQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080460.1 | NP_001073929.1 | 1 | 212 | ||
| XM_005247421.2 | XP_005247478.1 | 2 | 323 | |||
| XM_006713613.3 | XP_006713676.1 | 2 | 571 | |||
| XM_011512748.1 | XP_011511050.1 | 2 | 322 | |||
| XM_011512749.1 | XP_011511051.1 | 2 | 323 | |||

LRRIQ4
Genomic Map