siRNA Details
| Gene Symbol: GNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102386.2 | NP_001095856.1 | 5 | 633 | ||
| XM_011516124.2 | XP_011514426.1 | 5 | 1127 | |||
| GenBank mRNA | BC147016.1 | - | 5 | 633 | 9056944 | |
| BC147017.1 | - | 5 | 633 | 9056945 | ||

GNAT3
Genomic Map