siRNA Details
| Gene Symbol: MAGEB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271752.1 | NP_001258681.1 | 2 | 315 | ||
| GenBank mRNA | CR747535.1 | - | 2 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAGEB5| Gene Symbol: MAGEB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271752.1 | NP_001258681.1 | 2 | 315 | ||
| GenBank mRNA | CR747535.1 | - | 2 | 315 | ||