siRNA Details
| Gene Symbol: C6orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026736.1 | 5 | 472 | |||
| XM_006715088.3 | XP_006715151.1 | 5 | 666 | |||
| XM_011514572.2 | XP_011512874.1 | 4 | 462 | |||
| XM_011514575.2 | XP_011512877.1 | 5 | 648 | |||
| GenBank mRNA | BX280692.1 | - | 5 | 472 | ||

C6orf52
Genomic Map