siRNA Details
| Gene Symbol: NIPAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099287.1 | NP_001092757.1 | 6 | 3063 | ||
| NM_001172292.1 | NP_001165763.1 | 5 | 3006 | |||
| XM_011534552.1 | XP_011532854.1 | 6 | 2975 | |||
| GenBank mRNA | AF131815.1 | - | 1 | 1400 | ||
| AK026158.1 | - | 6 | 2867 | |||

NIPAL4
Genomic Map