siRNA Details
| Gene Symbol: PRAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282275.1 | NP_001269204.1 | 2 | 559 | ||
| NM_001282276.1 | NP_001269205.1 | 2 | 493 | |||
| XM_006721865.3 | XP_006721928.1 | 2 | 1175 | |||
| XM_011524747.2 | XP_011523049.1 | 2 | 485 | |||
| XM_011524748.1 | XP_011523050.1 | 2 | 430 | |||
| GenBank mRNA | AF532777.1 | - | 2 | 547 | ||
| BX106051.1 | - | 2 | 493 | |||

PRAC2
Genomic Map