siRNA Details
| Gene Symbol: LOC374890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135233.1 | 2 | 517 | |||
| GenBank mRNA | BC030765.2 | - | 2 | 517 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 LOC374890
LOC374890| Gene Symbol: LOC374890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135233.1 | 2 | 517 | |||
| GenBank mRNA | BC030765.2 | - | 2 | 517 | ||
 Back To Top
 Back To Top