siRNA Details
| Gene Symbol: EXOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018303.5 | NP_060773.3 | 3 | 388 | ||
| NR_073064.1 | 5 | 550 | ||||
| XM_017011018.1 | XP_016866507.1 | 4 | 434 | |||
| XM_017011019.1 | XP_016866508.1 | 4 | 596 | |||
| XM_017011020.1 | XP_016866509.1 | 4 | 456 | |||
| XM_017011021.1 | XP_016866510.1 | 3 | 692 | |||
| XM_017011022.1 | XP_016866511.1 | 3 | 388 | |||
| XM_017011023.1 | XP_016866512.1 | 3 | 388 | |||
| XM_017011024.1 | XP_016866513.1 | 4 | 434 | |||
| XM_017011025.1 | XP_016866514.1 | 3 | 388 | |||
| XM_017011026.1 | XP_016866515.1 | 3 | 388 | |||
| GenBank mRNA | AJ414403.1 | - | 3 | 312 | ||
| AK056564.1 | - | 3 | 356 | |||
| AK314628.1 | - | 3 | 388 | |||
| AL833213.1 | - | 3 | 334 | |||
| BC080542.1 | - | 5 | 520 | 5479496 | ||
| DA228027.1 | - | 3 | 388 | |||

EXOC2
Genomic Map