siRNA Details
| Gene Symbol: MROH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288814.1 | NP_001275743.1 | 36 | 4173 | ||
| NM_032450.2 | NP_115826.2 | 38 | 4313 | |||
| XM_011517270.2 | XP_011515572.1 | 38 | 4451 | |||
| XM_011517271.2 | XP_011515573.1 | 38 | 4450 | |||
| XM_011517272.2 | XP_011515574.1 | 37 | 4424 | |||
| XM_011517273.2 | XP_011515575.1 | 38 | 4449 | |||
| XM_011517274.2 | XP_011515576.1 | 37 | 4422 | |||
| XM_011517275.2 | XP_011515577.1 | 38 | 4449 | |||
| XM_011517276.2 | XP_011515578.1 | 38 | 4477 | |||
| XM_011517277.1 | XP_011515579.1 | 38 | 4313 | |||
| XM_011517278.1 | XP_011515580.1 | 37 | 4193 | |||
| XM_011517279.2 | XP_011515581.1 | 37 | 4357 | |||
| XM_011517281.1 | XP_011515583.1 | 37 | 4366 | |||
| XM_011517282.1 | XP_011515584.1 | 36 | 4291 | |||
| XM_011517283.2 | XP_011515585.1 | 38 | 4272 | |||
| XM_017013802.1 | XP_016869291.1 | 37 | 4422 | |||
| XM_017013803.1 | XP_016869292.1 | 37 | 4421 | |||
| XM_017013804.1 | XP_016869293.1 | 37 | 4286 | |||
| XM_017013805.1 | XP_016869294.1 | 36 | 4166 | |||
| XM_017013806.1 | XP_016869295.1 | 35 | 3928 | |||
| GenBank mRNA | AL834145.1 | - | 21 | 2571 | ||
| AL834309.1 | - | 37 | 4197 | |||
| BC017118.1 | - | 6 | 730 | |||

MROH1
Genomic Map