siRNA Details
| Gene Symbol: LOC105376351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136146.1 | 3 | 580 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376351| Gene Symbol: LOC105376351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136146.1 | 3 | 580 | |||