siRNA Details
| Gene Symbol: LINC00502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047467.2 | 4 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00502| Gene Symbol: LINC00502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047467.2 | 4 | 448 | |||