siRNA Details
| Gene Symbol: SAYSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304793.1 | NP_001291722.1 | 3 | 4827 | ||
| NM_018322.2 | NP_060792.1 | 2 | 420 | |||
| GenBank mRNA | AK001963.1 | - | 2 | 323 | ||
| AK124992.1 | - | 2 | 3081 | |||
| AL136839.1 | - | 2 | 375 | |||
| BC022007.2 | - | 2 | 344 | 4825803 | ||
| CR627244.1 | - | 3 | 4711 | |||
| DA977455.1 | - | 2 | 420 | |||

SAYSD1
Genomic Map