siRNA Details
| Gene Symbol: LOC101928617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135037.1 | 4 | 612 | |||
| GenBank mRNA | BC044741.1 | - | 4 | 612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928617| Gene Symbol: LOC101928617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135037.1 | 4 | 612 | |||
| GenBank mRNA | BC044741.1 | - | 4 | 612 | ||