siRNA Details
| Gene Symbol: TMEM233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005253881.3 | XP_005253938.1 | 3 | 2079 | ||
| XM_011538328.2 | XP_011536630.1 | 3 | 2080 | |||
| XM_011538329.2 | XP_011536631.1 | 3 | 2080 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM233| Gene Symbol: TMEM233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005253881.3 | XP_005253938.1 | 3 | 2079 | ||
| XM_011538328.2 | XP_011536630.1 | 3 | 2080 | |||
| XM_011538329.2 | XP_011536631.1 | 3 | 2080 | |||