siRNA Details
| Gene Symbol: LINC00398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047012.1 | 2 | 854 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00398| Gene Symbol: LINC00398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047012.1 | 2 | 854 | |||