siRNA Details
| Gene Symbol: LOC107984562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020896.1 | XP_016876385.1 | 2 | 713 | ||
| XM_017020897.1 | XP_016876386.1 | 3 | 298 | |||
| XM_017020898.1 | XP_016876387.1 | 3 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107984562| Gene Symbol: LOC107984562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020896.1 | XP_016876385.1 | 2 | 713 | ||
| XM_017020897.1 | XP_016876386.1 | 3 | 298 | |||
| XM_017020898.1 | XP_016876387.1 | 3 | 301 | |||