siRNA Details
| Gene Symbol: LINC00374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132367.1 | 3, 4 | 424 | |||
| GenBank mRNA | BG190276.1 | - | 3, 4 | 394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00374| Gene Symbol: LINC00374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132367.1 | 3, 4 | 424 | |||
| GenBank mRNA | BG190276.1 | - | 3, 4 | 394 | ||