siRNA Details
Gene Symbol: LINC01078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132375.1 | 3 | 252 | |||
GenBank mRNA | AA442466.1 | - | 3 | 257 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132375.1 | 3 | 252 | |||
GenBank mRNA | AA442466.1 | - | 3 | 257 |