siRNA Details
| Gene Symbol: LOC101929613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110947.1 | 3, 4 | 95 | |||
| NR_110948.1 | 3, 4 | 95 | ||||
| GenBank mRNA | HM587423.1 | - | 3, 4 | 95 | ||
| HM587424.1 | - | 3, 4 | 95 | |||
| HM587425.1 | - | 3, 4 | 96 | |||

LOC101929613
Genomic Map