siRNA Details
| Gene Symbol: MIR193BHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132983.1 | 2 | 640 | |||
| NR_132984.1 | 2 | 362 | ||||
| GenBank mRNA | CD675241.1 | - | 2 | 640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR193BHG| Gene Symbol: MIR193BHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132983.1 | 2 | 640 | |||
| NR_132984.1 | 2 | 362 | ||||
| GenBank mRNA | CD675241.1 | - | 2 | 640 | ||